Learn More
Raybiotech Inc C-Series Porcine Cytokine Array C1 Kit.
Supplier:  Raybiotech Inc AAPCYT12
Encompass_Preferred
Detects 48 Porcine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Angiopoietin-1, CCL3L1, Decorin, Eotaxin-1, Erythropoietin, FGF-21, Galectin-9, GASP-1, GM-CSF, IFN alpha, IFN-beta, IFN-gamma, IGF-2, IGFBP-5, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-17F, IL-18, IL-21, IL-22, IL-28B, IL-4, IL-6, IL-8, Insulin, MIF, MIG, MIP-1 beta, NCAM-1, Osteoprotegerin, PDGF-BB, PECAM-1, PLGF-2, RANTES, SCF, TGF alpha, TGF beta 1, TIMP-1, TIMP-2, TNF alpha, TWEAK R, VEGF-A
Catalog No. 50-194-8752