Learn More
Raybiotech Inc G-Series Porcine Cytokine Array 2 Kit.
Catalog No. 501950486
Encompass_Preferred
Semi-quantitative measurement of 10 porcine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CCL3L1, IFN alpha, IL-1 alpha (IL-1 F1), IL-1 Ra (IL-1 F3), IL-13, IL-17A, IL-18, MIG (CXCL9), MIP-1 beta (CCL4), PECAM-1 (CD31)
Catalog No. 50-195-0486 Supplier Raybiotech Inc Supplier No. GSPCYT21
Semi-quantitative measurement of 10 porcine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CCL3L1, IFN alpha, IL-1 alpha (IL-1 F1), IL-1 Ra (IL-1 F3), IL-13, IL-17A, IL-18, MIG (CXCL9), MIP-1 beta (CCL4), PECAM-1 (CD31)