Learn More
Raybiotech Inc G-Series Porcine Cytokine Array 3 Kit.
Catalog No. 501950489
Encompass_Preferred
Semi-quantitative measurement of 10 porcine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Decorin, GASP-1 (WFIKKNRP), IGFBP-5, IL-15, IL-22, Insulin, IP-10 (CXCL10), MCP-1(CCL2), NCAM-1 (CD56), TWEAK R (TNFRSF12)
Catalog No. 50-195-0489 Supplier Raybiotech Inc Supplier No. GSPCYT32
Semi-quantitative measurement of 10 porcine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Decorin, GASP-1 (WFIKKNRP), IGFBP-5, IL-15, IL-22, Insulin, IP-10 (CXCL10), MCP-1(CCL2), NCAM-1 (CD56), TWEAK R (TNFRSF12)