Learn More
Raybiotech Inc G-Series Porcine Cytokine Array 4 Kit.
Supplier:  Raybiotech Inc GSPCYT41
Encompass_Preferred
Semi-quantitative measurement of 10 porcine cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Angiopoietin-1, IL-17F, MIF, Osteoprotegerin (TNFRSF11B), PDGF-BB, RANTES (CCL5), TGF alpha, TIMP-1, TIMP-2, VEGF-A
Catalog No. 50-195-0490