Learn More
Raybiotech Inc G-Series Porcine Cytokine Array 5 Kit.
Supplier:  Raybiotech Inc GSPCYT52
Encompass_Preferred
Detects 10 Porcine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Eotaxin-1, Erythropoietin, FGF-21, Galectin-9, IFN beta, IGF-2, IL-21, IL-28B, PlGF-2, SCF
Catalog No. 50-195-0493