Learn More
Medchemexpress LLC HY-P0088 1mg Medchemexpress, Porcine dynorphin A(1-13) CAS:72957-38-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP00881MG
Encompass_Preferred
Medchemexpress, HY-P0088 1mg Porcine dynorphin A(1-13) CAS:72957-38-1 Porcine dynorphin A (1-13) is a potent, endogenous κ opioid receptor agonist and is antinociceptive at physiological concentrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4015