Learn More
Raybiotech Inc Porcine FGF-19 ELISA Kit
Supplier:  Raybiotech Inc ELPFGF191
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Porcine FGF-19. Other species not determined.
Catalog No. 50-195-0183