Learn More
Raybiotech Inc Porcine GM-CSF ELISA Kit
Supplier:  Raybiotech Inc ELPGMCSF1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 72 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: porcine IL-1 beta, IL-4, IL-6, IL-8, IL-10, IL-12, IFN-gamma, TGF beta 1, TNF alpha
Catalog No. 50-195-0184