Learn More
Alpco Ltd Porcine Haptoglobin ELISA 96 Wells 5 L
SDP
Supplier:  Alpco Ltd 41HAPPOE01
Encompass
The Porcine Haptoglobin ELISA is for the determination of Haptoglobin in Serum or Plasma of pigs. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-922-272