Learn More
Raybiotech Inc Porcine IFN alpha ELISA Kit
Supplier:  Raybiotech Inc ELPIFNA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 36 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects porcine IFN-alpha. Other species not determined.
Catalog No. 50-195-0185