Learn More
Abcam Porcine IFN-alpha ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2732131X96TESTS
Encompass_Preferred
Porcine IFN-alpha ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Porcine IFN-alpha in Pig in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1894200