Learn More
Raybiotech Inc Porcine IL-1 beta (IL-1 F2) ELISA Kit
Supplier:  Raybiotech Inc ELPIL1B1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 6 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Porcine IL-1 beta.
Catalog No. 50-195-0195