Learn More
Raybiotech Inc Porcine IL-15 ELISA Kit
Supplier:  Raybiotech Inc ELPIL151
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 7.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Porcine IL-15.
Catalog No. 50-195-0190