Learn More
Raybiotech Inc Porcine IL-17A ELISA Kit
Supplier:  Raybiotech Inc ELPIL17A1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 14 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair also can recognize bovine and equine IL-17A.
Catalog No. 50-195-0191