Learn More
Raybiotech Inc Porcine IL-18 ELISA Kit
Supplier:  Raybiotech Inc ELPIL181
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 80 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: porcine CCL3L1, IFN alpha, IL-1 alpha, IL-1R alpha, IL-13, IL-17A, MIG, MIP-1 beta, PECAM-1
Catalog No. 50-195-0193