Learn More
Raybiotech Inc Porcine IL-22 ELISA Kit
Supplier:  Raybiotech Inc ELPIL221
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 20 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects porcine IL-22. Other species not determined.
Catalog No. 50-195-0196