Learn More
Raybiotech Inc C-Series Porcine Inflammation Array C1 Kit.
Catalog No. 501948758
Encompass_Preferred
Detects 26 Porcine Inflammation factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Eotaxin-1, GM-CSF, IFN alpha, IFN-beta, IFN-gamma, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-17F, IL-18, IL-21, IL-22, IL-28B, IL-4, IL-6, IL-8, MIF, MIG, MIP-1 beta, RANTES, TNF alpha
Catalog No. 50-194-8758 Supplier Raybiotech Inc Supplier No. AAPINF12
Detects 26 Porcine Inflammation factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Eotaxin-1, GM-CSF, IFN alpha, IFN-beta, IFN-gamma, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-17F, IL-18, IL-21, IL-22, IL-28B, IL-4, IL-6, IL-8, MIF, MIG, MIP-1 beta, RANTES, TNF alpha