Learn More
Raybiotech Inc Porcine Insulin ELISA Kit
Supplier:  Raybiotech Inc ELPINSULIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 4 uIU/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects porcine, mouse, human, rat, and bovine Insulin.
Catalog No. 50-195-0200