Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Porcine Insulin ELISA Kit

Catalog No. ES16RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES16RBX5 5 x 96 Tests
ES16RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES16RBX5 Supplier Invitrogen™ Supplier No. ES16RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Porcine Insulin quantitates porcine Insulin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine Insulin.

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01308, P01315, P01317, P01322, P01323, P01325, P01326
Assay Range 4-300 μIU/mL
Assay Sensitivity 4 μIU/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human, Mouse, Rat, Bovine, Pig
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulinAA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin;
Gene ID (Entrez) 16333, 16334, 24505, 24506, 280829, 3630, 397415
Gene Symbol INS, Ins1, Ins2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Insulin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.