Learn More
Mercodia Inc Porcine Insulin ELISA, Pig, Serum Plasma, 96 Wells
SDP
Supplier:  Mercodia Inc 10120001
Encompass_Preferred
The Mercodia Porcine Insulin ELISA is a high quality, species-optimized enzyme immunoassay for the quantification of porcine insulin in serum, plasma, or cell culture media. Specifically optimized for porcine sample matrix. No cross-reactivity to proinsulin or c-peptide. Highly sensitive.
Catalog No. NC9623145