Learn More
Bioworld PORCINE INTERLEUKIN-2 (RECO
Supplier:  Bioworld 220606341
Encompass_Preferred
PORCINE INTERLEUKIN-2 (RECOMBINANT): Recombinant Porcine Interleukin-2 (IL-2) Source: E.Coli Description: Recombinant Porcine IL-2 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 134 amino acids and havi...
Catalog No. 50-253-733