Learn More
Bioworld PORCINE INTERLEUKIN-6 (RECO
Supplier:  Bioworld 220606361
Encompass_Preferred
PORCINE INTERLEUKIN-6 (RECOMBINANT): Recombinant Porcine Interleukin-6 (IL-6) Source: E.Coli Background : Cytokines of the IL6/GCSF/MGF family are glycoproteins of about 170 to 180 amino acid residues that contains four conserved cys...
Catalog No. 50-253-735