Learn More
Raybiotech Inc Porcine MIF ELISA Kit
Supplier:  Raybiotech Inc ELPMIF1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 16 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: porcine Angiopoietin-1, IL-17F, Osteoprotegerin, PDGF-BB, RANTES, TGF-alpha, TIMP-1, TIMP-2, VEGF-A.
Catalog No. 50-195-0202