Learn More
Raybiotech Inc Porcine Osteoprotegerin (TNFRSF11B) ELISA Kit
Supplier:  Raybiotech Inc ELPOPG1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Porcine Osteoprotegerin (TNFRSF11B)
Catalog No. 50-195-0203