Learn More
Innovative Research Inc Porcine Red Blood Cells Packed 10% 15ml
Catalog No. 502034870
Encompass_Preferred
Porcine Red Blood Cells from Innovative Research are manufactured using whole blood collections from Pigs. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 10% suspension in Alsever's Solution. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4870 Supplier Innovative Research Inc Supplier No. IPCRBC10P15ML
Porcine Red Blood Cells from Innovative Research are manufactured using whole blood collections from Pigs. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 10% suspension in Alsever's Solution. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.