Learn More
Medchemexpress LLC Porcine serum albumin | 9048-46-8 | 99.9% | 5 MG
SDP
Catalog No. 5000425314
Encompass_Preferred
Porcine serum albumin is a serum albumin protein with a flexible C-terminal region, functioning as both a pork allergen and a biomarker for detection. It offers potential in preventing and treating pork allergy, developing hypoallergenic pork products, and can be conjugated with polyoxazoline for use as an artificial plasma expander in dogs, exhibiting moderate colloid osmotic pressure and good blood cell compatibility.
  • Functions as a pork allergen
  • Acts as a biomarker for detecting pork
  • Modifications may be used for preventing and treating pork allergy
  • Modifications may be used for producing hypoallergenic pork products
  • Can be conjugated with polyoxazoline to serve as an artificial plasma expander for dogs
  • Exhibits moderately high colloid osmotic pressure
  • Shows good blood cell compatibility
Catalog No. 50-004-25314 Supplier Medchemexpress LLC Supplier No. HYNP002E5MG
Porcine serum albumin is a serum albumin protein with a flexible C-terminal region, functioning as both a pork allergen and a biomarker for detection. It offers potential in preventing and treating pork allergy, developing hypoallergenic pork products, and can be conjugated with polyoxazoline for use as an artificial plasma expander in dogs, exhibiting moderate colloid osmotic pressure and good blood cell compatibility.
  • Functions as a pork allergen
  • Acts as a biomarker for detecting pork
  • Modifications may be used for preventing and treating pork allergy
  • Modifications may be used for producing hypoallergenic pork products
  • Can be conjugated with polyoxazoline to serve as an artificial plasma expander for dogs
  • Exhibits moderately high colloid osmotic pressure
  • Shows good blood cell compatibility