Learn More
Sigma Aldrich Fine Chemicals Biosciences Porcine srum origin s500ML
Catalog No. 502900323
Encompass
The porcine serum comprises serum amyloid A (SAA) C-reactive protein (CRP) haptoglobin (Hp) and pig major acute phase protein (pig-MAP).
Catalog No. 50-290-0323 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P9783500ML
The porcine serum comprises serum amyloid A (SAA) C-reactive protein (CRP) haptoglobin (Hp) and pig major acute phase protein (pig-MAP).