Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Porcine TIMP-1 ELISA Kit

Catalog No. ES22RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES22RBX5 5 x 96 Tests
ES22RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES22RBX5 Supplier Invitrogen™ Supplier No. ES22RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Porcine TIMP-1 quantitates porcine TIMP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine TIMP-1.

TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01033, P35624
Assay Range 18-4000 pg/mL
Assay Sensitivity 18 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human, Pig
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1;
Gene ID (Entrez) 396862, 7076
Gene Symbol TIMP1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target TIMP1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.