Learn More
Raybiotech Inc Porcine TIMP-2 ELISA Kit
Supplier:  Raybiotech Inc ELPTIMP21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 28 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects porcine and human TIMP-2. Other species not determined.
Catalog No. 50-195-0208