Learn More
Raybiotech Inc Porcine TNF alpha ELISA Kit
Supplier:  Raybiotech Inc ELPTNFA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 20 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Porcine TNF-alpha.
Catalog No. 50-195-0209