Learn More
Aviva Systems Biology Corp Porcn Antibody - middle region (ARP49763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Porcn. It was validated on Western Blot. . Alternate names: 2410004O13Rik, AW045557, DXHXS7465e, Mporc, Mporc-a, Mporc-b, Mporc-c, Mporc-d, Ppn, mMg61, porcApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-447