Learn More
Aviva Systems Biology Corp PORCN Antibody - N-terminal region (ARP49762_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49762_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PORCN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PORCN. Alternate names: DHOF, FODH, MG61, MGC29687, PORC, PPN, porApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53601