Learn More
Medchemexpress LLC HY-111472 10mg Medchemexpress, Porcn-IN-1 CAS:2036044-77-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11147210MG
Encompass_Preferred
Medchemexpress, HY-111472 10mg Porcn-IN-1 CAS:2036044-77-4 Porcn-IN-1 is potent porcupine inhibitor with an IC50 of 0.5±0.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8878