Learn More
Medchemexpress LLC Porcn-IN-1 | 2036044-77-4 | 99.6% | 410.44 | 25 MG
SDP
Catalog No. 5000346708
Encompass_Preferred
Porcn-IN-1 is a potent porcupine inhibitor with an IC50 of 0.5±0.2 nM. It is as potent as the clinical compound LGK974 in a cell-based STF reporter gene assay and potently inhibits the secretion of Wnt3A, confirming it as a porcupine inhibitor. This product is for research use only and not intended for human use.
  • Potent porcupine inhibitor
  • Exhibits moderate clearance in human and rat liver microsomes
  • Shows high clearance in mouse microsomes
  • Soluble in DMSO at ≥ 100 mg/mL
Catalog No. 50-003-46708 Supplier Medchemexpress LLC Supplier No. 25MGHY111472
Porcn-IN-1 is a potent porcupine inhibitor with an IC50 of 0.5±0.2 nM. It is as potent as the clinical compound LGK974 in a cell-based STF reporter gene assay and potently inhibits the secretion of Wnt3A, confirming it as a porcupine inhibitor. This product is for research use only and not intended for human use.
  • Potent porcupine inhibitor
  • Exhibits moderate clearance in human and rat liver microsomes
  • Shows high clearance in mouse microsomes
  • Soluble in DMSO at ≥ 100 mg/mL