Learn More
Cayman Chemical PorcupIn-IN-1 10mg
SDP
Catalog No. 502936941
Encompass_Preferred
An inhibitor of the Wnt and hedgehog signaling pathways (IC50s = 0.5 and 71 nM, respectively, in cell-based reporter assays); inhibits secretion of Wnt3A from Wnt3A-transfected HEK293T cells at 0.1 μM, indicating inhibition of porcupine
Catalog No. 50-293-6941 Supplier Cayman Chemical Supplier No. 2583410 MG
An inhibitor of the Wnt and hedgehog signaling pathways (IC50s = 0.5 and 71 nM, respectively, in cell-based reporter assays); inhibits secretion of Wnt3A from Wnt3A-transfected HEK293T cells at 0.1 μM, indicating inhibition of porcupine