Learn More
Selleck Chemical LLC Poricoic acid A-E0204-1MG
SDP
Catalog No. 503138359
Encompass_Preferred
Poricoic acid A is isolated from the surface layer of Poria cocos Poricoic acid A activates AMPK to attenuate fibroblast activation and abnormal extracellular matrix remodelling in renal fibrosis Poricoic acid A also is a modulator of tryptophan hydroxylase-1 (TPH-1)
Catalog No. 50-313-8359 Supplier Selleck Chemical LLC Supplier No. E02041MG
Poricoic acid A is isolated from the surface layer of Poria cocos Poricoic acid A activates AMPK to attenuate fibroblast activation and abnormal extracellular matrix remodelling in renal fibrosis Poricoic acid A also is a modulator of tryptophan hydroxylase-1 (TPH-1)