Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ HQ 50 μm Strong Anion Exchange Resin

Catalog No. p-6927060 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PI82077 10 mL
PI82078 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PI82077 Supplier Thermo Scientific™ Supplier No. 82077
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For Research Use Only. Not for use in diagnostic procedures.

POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that enables >140 mg/mL dynamic binding capacity while delivering exceptional separation performance

POROS™ 50-micron resins are available for low pressure applications, including large scale downstream bioprocessing. POROS HQ is based on a quaternized polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography™ resin designed for the separation and purification of biomolecules.

  • Higher productivity—high throughput and high dynamic capacity associated with Perfusion Chromatography
  • Chemical stability—allows aggressive cleaning and sanitization
  • Enhanced biomolecule access—provided via large pores, ranging between 500-10000 Å
  • Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
  • Develop better separations methods in a shorter time frame—the speed of Perfusion Chromatography technology reduces weeks of experimentation to only a few hours of work, so you have plenty of time to explore all the variables of your separation

Reduce time-consuming sample prep
You can replace many sample preparation steps with high-speed Perfusion Chromatography technology. For instance, dialysis of large volumes of material can be replaced by high flow rate processing of the dilute sample. Elution in a small volume of new buffer accomplishes both buffer exchange and concentration.

Create novel assays for more efficient analysis
Perfusion Chromatography technology is not limited to standard modes of separation. Both enzymes and affinity ligands can be immobilized on POROS media. By combining rapid on-column protein digestions with rapid on-column immunoassays and chromatographic separations, you can create entirely new assays with unlimited potential.

High capacity, high resolution, high speed
In contrast to conventional chromatography media, POROS Perfusion Chromatography resin particles are engineered to have two discreet classes of pores. Large throughpores allow convection flow to occur through the particles themselves, quickly carrying sample molecules to short diffusive pores inside. By reducing the distance over which diffusion needs to occur, the time required for sample molecules to interact with interior binding sites is also reduced. Diffusion is no longer limiting and flow rates can be dramatically increased—without compromising resolution or capacity. Separations can be achieved at 1,000 to 5,000 cm/hr compared to 50 to 360 cm/hr for conventional resins

TRUSTED_SUSTAINABILITY

Specifications

Type HQ Strong Anion Exchange Resin
Standard Components Quaternized Polyethyleneimine
For Use With (Application) Separation and Purification of Biomolecules
Quantity 10 mL
Volume (Metric) 10 mL
Content And Storage Store in refrigerator (2–8°C).
Product Line POROS
Format Poros Bead
Stationary Phase Quaternized Polyethyleneimine
Purification Target Protein
Shipping Condition Room Temperature
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.