Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ Benzyl Ultra RoboColumn™ Pre-packed columns
GREENER_CHOICE

Catalog No. A34816 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Strip, 8 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A34816 1 Strip, 8 Columns
A34815 1 Strip, 8 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A34816 Supplier Thermo Scientific™ Supplier No. A34816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific™ RoboColumn™ product is pre-packed with POROS Benzyl Ultra Hydrophobic Interaction Chromatography (HIC) Resin that is well suited for aggregate removal from monoclonal antibody monomer in flow-through mode at low salt concentrations in large scale downstream bioprocessing.

This RoboColumn™ product is pre-packed with POROS Benzyl Ultra Hydrophobic Interaction Chromatography (HIC) Resin (Cat. No. A32564) that is well suited for aggregate removal from monoclonal antibody monomer in flow-through mode at low salt concentrations in large scale downstream bioprocessing.

RoboColumn™ products are small chromatography columns that are provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.Features of POROS Benzyl Ultra HIC Resin include:

  • Superior resolution capability while maintaining excellent pressure-flow properties, achieved by combining the high-flow characteristics of a POROS base bead with a 50-micron particle size. Higher flow rates help improve productivity.
  • Unique selectivity due to the novel hydrophobic ligand, allowing for purification of a wide range of biomolecules, including therapeutic proteins, antibody fragments, antibody drug conjugates, and enzymes
  • High capacity, reducing the process volume significantly for subsequent steps
  • A rigid polymeric bead with covalent surface chemistry for easier handling and packing with superior physical and chemical stability, allowing cleaning with various clean-in-place (CIP) solutions such as sodium hydroxide and enabling a more robust downstream purification process

These features of POROS Benzyl Ultra HIC Resin are a major improvement to the performance of traditional HIC resins, setting the standard for hydrophobic interaction chromatography.

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Pore Size Not Applicable
Stationary Phase High-Density Benzyl/Aromatic
Column Capacity 600 μL
Length (Metric) 3 cm
Quantity 1 Strip, 8 Columns
Particle Size 50 μm
For Use With (Equipment) Robotic Fluid Handlers
Type HIC Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 30°C. Do not freeze.
Product Line POROS
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Shipping Condition Ambient
Column Format Pre-packed Robo Column
Bed Volume 600 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.