Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ AAVX Affinity Resin
GREENER_CHOICE

Catalog No. A36743 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
50 mL
250 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A36743 1 L
A36739 10 mL
A36740 25 mL
A36741 50 mL
A36742 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A36743 Supplier Thermo Scientific™ Supplier No. A36743

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Thermo Scientific™ POROS™ CaptureSelect™ AAVX Affinity Resin is engineered to address the high selectivity and capacity requirements for large-scale downstream purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications.

POROS CaptureSelect AAVX Affinity Resin is engineered to address the high selectivity and capacity requirements for large-scale downstream purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. POROS CaptureSelect AAVX resin has demonstrated binding reactivity towards a set of AAV serotypes that includes AAV1 to AAV8, and AAVrh10 as well as synthetic serotypes.

Features of POROS CaptureSelect AAVX resin include:

  • One-step AAV purification from crude material with high purity and yield
  • High specificity and capacity, maximizing yield by significantly reducing process steps
  • Superior capacity maintained at high flow rates
  • Excellent scalability and easier handling and packing due to rigid polymeric bead
  • Non-animal derived

POROS CaptureSelect AAVX Affinity Resin enables a robust, fast, and platformable purification process for wide-ranging AAV serotypes with excellent purity and yield obtained in one step. This resin comes with complete technical and regulatory support.

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: CaptureSelect AAVX affinity ligand
  • Binding capacity: >1 x 1013 genome copies/mL of resin
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1450 psi; 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store in refrigerator at 2°C to 8°C.
Shipping Condition Wet Ice
Particle Size 50 μm
Form Liquid Suspension
Product Line POROS
Quantity 1 L
Type Resin
Column Type Affinity
Binding Capacity >1 x 1013 Genome Copies/mL of Resin

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.