Learn More
Thermo Scientific™ POROS™ CaptureSelect™ AdV5 Affinity Matrix

Description
POROS CaptureSelect AdV5 Affinity Matrix was designed specifically for the purification and/or scavenging of recombinant hexon capsomeres of Adenovirus serotype 5 (AdV5) particles, as well as free hexon proteins thereof, for high purity in a single step. This affinity resin specifically binds AdV5 virus particles and enables a robust, fast, and efficient purification for research applications with excellent purity and yield in one step.
Features of this affinity matrix include:
- One-step purification of virus particles from crude material with high purity and yield
- High specificity, capacity, and retained activity
- Excellent scalability
- Non-animal derived
- High flow rates
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: cross-linked poly(styrene-divinylbenzene)
- Average particle size: 50 μm
- Ligand: POROS CaptureSelect AdV5 affinity ligand
- Ligand coupling method: epoxide
- Binding capacity: >1 x 1011 genome copies/mL of resin
- Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
- Mechanical resistance: 100 bar (1450 psi, 10 MPa)
- Formulation buffer: 20% (v/v) ethanol
Specifications
Specifications
| Binding Capacity | >1 x 1011 genome copies/mL of resin |
| Matrix | Cross-linked poly(styrene-divinylbenzene) |
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Particle Size | 50 μm |
| Form | Liquid Suspension |
| Product Line | CaptureSelect, POROS |
| Quantity | 50 mL |
| Type | Affinity Matrix |
| Ligand Type | AdV5 affinity ligand |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.