Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ BacuClear Affinity Matrix
GREENER_CHOICE

Catalog No. 2813742005
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-137-42005 5 mL
28-137-42010 10 mL
28-137-42050 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-137-42005 Supplier Thermo Scientific™ Supplier No. 2813742005
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ POROS™ CaptureSelect™ BacuClear Affinity Matrix is specifically designed for the removal of baculovirus contaminants when using baculovirus expression systems (BEVS) for the production of recombinant biomolecules.

POROS CaptureSelect BacuClear Affinity Matrix is specifically designed for the removal of baculovirus contaminants when using baculovirus expression systems (BEVS) for the production of recombinant biomolecules. It is particularly useful when expressing virus and virus-like particles (VLPs), which are similar in size and envelope structure to baculovirus, making discrimination and separation from the co-expressed baculovirus contaminants particularly challenging. The baculovirus-specific CaptureSelect affinity ligand was developed to address this challenge and is coupled to the high-performing POROS matrix. The large pore structure of the POROS beads enables efficient purification of larger biomolecules.

To remove baculovirus impurities, the affinity matrix is operated in flow-through mode. As described by Moleirinho et. al. (1), successful removal of baculovirus particles from influenza VLPs, HCV VLPs, and AAVs was achieved with the affinity ligand identified (BV-2) coupled to the POROS matrix. A dynamic binding capacity for baculovirus particles of 1.6 x 1010 VG/mL affinity matrix was demonstrated with baculovirus removals observed above 70% and VLP recovery yields above 60%.

Features of this affinity matrix include:

  • One-step removal of baculovirus virus particles from virus and VLP containing samples, in flow-through mode
  • Enables a highly selective polishing step for baculovirus removal
  • High specificity, capacity, and retained activity
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect BacuClear affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: estimated >1 x 1010 VG/mL of resin
  • Regeneration conditions: acidic stripping condition with 0.5 M acetic acid, pH 2
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 20% (v/v) ethanol

1. M.G. Moleirinho, R.P. Fernandes, S.B. Carvalho, S. Bezemer, F. Detmers, P Hermans, R.J.S. Silva, P.M. Alves, M.J.T. Carrondo, C. Peixoto. Baculovirus affinity removal in viral-based bioprocesses. Sep. Purif. Technol. 241 (2020)

Specifications

Binding Capacity >1 x 1010 VG/mL of resin (estimated)
Formulation 50% slurry suspension in 20% ethanol
Matrix Cross-linked poly(styrene-divinylbenzene)
Content And Storage Store in refrigerator (2°C to 8°C). Protect from daylight
Max. Operating Pressure 100 bar
Shipping Condition Ships at 2°C to 8°C
Particle Size 50 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type BacuClear affinity ligand
Stationary Phase POROS bead
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.