Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ IgM-XL Affinity Matrix
GREENER_CHOICE

Catalog No. p-9658000
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-128-92005 5 mL
28-128-92010 10 mL
28-128-92050 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 28-128-92005 Supplier Thermo Scientific™ Supplier No. 2812892005
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ POROS™ CaptureSelect™ IgM-XL Affinity Matrix is intended for the purification of human IgM, with no binding to other immunoglobulins.

POROS CaptureSelect IgM-XL Affinity Matrix is intended for the purification of human IgM, with no binding to other immunoglobulins.

Features of this affinity matrix include:

  • IgM purification from plasma and serum
  • Purification of monoclonal IgM from cell cultures
  • Efficient purification of IgM with high yields and retained activity
  • Excellent scalability
  • Non-animal derived
  • High flow rates
  • Cross-reactivity towards mouse and rat IgM

POROS CaptureSelect IgM-XL Affinity Matrix binds to the Fc part of IgM. The high selectivity and yields obtained using this affinity matrix enable a robust, fast, and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (animal origin-free).

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect IgM-XL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >5 g/L resin (determined with polyclonal human IgM from serum)
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Stationary Phase 20% Ethanol
Product Line CaptureSelect
Type Affinity Matrix
Quantity 5 mL
Form Resin Slurry
Particle Size 50 μm
Binding Capacity >5 g/L
Ligand Type IgM-XL affinity ligand
Matrix Cross-linked poly(styrene-divinylbenzene)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.