Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ Ethyl RoboColumn™ Pre-packed columns
GREENER_CHOICE

Catalog No. A34810
Change view
Click to view available options
Quantity:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A34810 200 μL
A34812 600 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A34810 Supplier Thermo Scientific™ Supplier No. A34810
Only null left
Add to Cart
Add to Cart

This Thermo Scientific™ RoboColumn™ product is pre-packed with POROS Ethyl Hydrophobic Interaction Chromatography (HIC) Resin and suitable for impurity removal during large-scale downstream purification of biomolecules, primarily for bind/elute operations

This RoboColumn™ product is pre-packed with POROS Ethyl Hydrophobic Interaction Chromatography (HIC) Resin (Cat. No. A32552) and suitable for impurity removal during large-scale downstream purification of biomolecules, primarily for bind/elute operations

RoboColumn™ products are small chromatography columns that are provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.

Features of POROS Ethyl HIC Resin include:

  • Superior resolution capability while maintaining excellent pressure-flow properties, achieved by combining the high-flow characteristics of a POROS base bead with a 50-micron particle size. Higher flow rates help improve productivity.
  • Unique selectivity due to the novel hydrophobic ligand, allowing for purification of a wide range of biomolecules, including therapeutic proteins, antibody fragments, antibody drug conjugates, and enzymes
  • High capacity, reducing the process volume significantly for subsequent steps
  • A rigid polymeric bead with covalent surface chemistry for easier handling and packing with superior physical and chemical stability, allowing cleaning with various clean-in-place (CIP) solutions such as sodium hydroxide and enabling a more robust downstream purification process

These features of POROS Ethyl HIC Resin are a major improvement to the performance of traditional HIC resins, setting the standard for hydrophobic interaction chromatography.

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Product Type Pre-packed Column
Quantity 200 μL
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Product Line POROS
Stationary Phase Novel Ethyl
Capacity (Metric) 200 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.