Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ MabCapture™ A/G Select Resin

Catalog No. p-7180365
Change view
Click to view available options
Quantity:
5 mL
15 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI82085 5 mL
PI82086 15 mL
PI82087 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI82085 Supplier Thermo Scientific™ Supplier No. 82085
Only null left
Add to Cart
Add to Cart

MabCapture A/G Select allows antibody purification with high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates

POROS™ MabCapture™ A/G Select Resin is a new high-throughput Protein A/G chromatography resin manufactured using our own Protein A/G ligand to enable high-throughput purification performance. This resin allows antibody purification with high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates where other chromatography media products cannot operate.

POROS MabCapture A/G Select Resin provides the most versatile combination of chromatographic features for high yield and high purity purification of whole IgG from a variety of mammalian serum samples. It allows antibody purification with high dynamic binding performance at low flow rates and maintains superior binding capacity at high linear velocities.

  • Maintain high dynamic binding capacity at linear flow rates over 700 cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates, or both) due to the improvements made to the antibody binding efficiency
  • Obtain the highest dynamic binding capacity for antibodies vs. conventional agarose- or silica-based Protein A or Protein G resins
  • Purify polyclonal IgG from nearly any mammalian species
  • Sanitize and depyrogenate with standard agents used for other chromatography products
  • Recognize major increases to process productivity and reduce the downstream bottleneck

Protein A/G is a chimeric genetically-engineered protein that combines the five IgG binding domains of Protein A and two IgG binding domains of Protein G. Protein A/G is not as pH-dependent as Protein A alone, but otherwise has the additive properties of Protein A and G. Protein A/G binds nearly all isotypes and mammalian species of IgG from serum, ascites fluid, cell culture supernatant and other antibody samples, thereby enabling effective antibody purification.

The development of humanized monoclonal antibodies has continued to fuel the growth of the biotherapeutic market. Addressing the therapeutic demand for monoclonal antibodies has led to major improvements to the cell-based expression systems that are used to manufacture these proteins. Consequently, these improvements have moved the processing bottleneck from upstream cell expression to downstream purification. POROS MabCapture A/G Select chromatography resin offers the capacity, efficiency, and flexibility needed to make improvements to the downstream purification platform, resulting in major increases to process productivity and antibody manufacturing.

TRUSTED_SUSTAINABILITY

Specifications

Type A/G Select Resin
Standard Components Protein A/G
For Use With (Application) IgG Purification
Quantity 5 mL
Content And Storage Store in refrigerator (2–8°C).
Product Line POROS
Format Poros Bead
Stationary Phase Protein A/G
Purification Target IgG Antibodies, Ig Antibodies
Shipping Condition Wet Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.