Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ MabCapture™ A Select Resin

Catalog No. PI82080 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
5 mL
15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PI82080 15 mL
PI82079 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PI82080 Supplier Thermo Scientific™ Supplier No. 82080

missing translation for 'validMainframeMsg'

For Research Use Only. Not for use in diagnostic procedures.

MabCapture A Select a monoclonal antibody purification resin provides high dynamic binding at low linear flow rates and maintains superior binding capacity at high linear flow rates

POROS™ MabCapture™ A Select Resin is a new high-throughput, lower-priced Protein A chromatography resin, combining past improvements to MabCapture A with our own Protein A ligand, enabling the same high-throughput purification performance at a significant reduction in price over most Protein A chromatography resins. These advances provide a monoclonal antibody purification resin that demonstrates high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates where other chromatography media products cannot operate.

  • Maintain high dynamic binding capacity at linear flow rates over 700 cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates, or both) due to the improvements made to the antibody binding efficiency
  • Obtain the highest dynamic binding capacity for antibodies vs. conventional agarose- or silica-based Protein A resins
  • Purify for polyclonal IgG from human, rabbit, monkey, mouse, pig, dog, or cat serum
  • Clean and sanitize with standard sodium hydroxide agents used for other chromatography products
  • Recognize major increases to process productivity and reduce the downstream bottleneck

The Protein A molecule contains five high-affinity binding sites capable of binding to the Fc portion of antibodies. Compared to Protein G, Protein A provides the highest binding capacity for subclasses of IgG from rabbits, pigs, dogs, and cats. Protein A also has higher binding capacity for human IgG, except for IgG3, which it binds weakly. Protein A also binds weakly to mouse IgG1 and is not recommended for most applications with that antibody isotype. The Protein A molecule also has greater stability than Protein G. Protein A is heat-stable and retains its native conformation when exposed to denaturing reagents such as 4M urea, 4M thiocyanate, and 6M guanidine hydrochloride.

The development of humanized monoclonal antibodies has continued to fuel the growth of the biotherapeutic market. Addressing the therapeutic demand for monoclonal antibodies has led to major improvements to the cell-based expression systems that are used to manufacture these proteins. >

Consequently, these improvements have moved the processing bottleneck from upstream cell expression to downstream purification. Protein A chromatography, which is used as the primary capture chromatography step in most antibody process platforms, is generally regarded as the major process bottleneck due to the slow flow rates and long residence times required for the effective operation of conventional Protein A chromatography resin. POROS MabCapture Select chromatography resin offers the capacity, efficiency, and flexibility needed to make improvements to the downstream purification platform, resulting in major increases to process productivity and antibody manufacturing

TRUSTED_SUSTAINABILITY

Specifications

Type A Select Resin
Standard Components Protein A
For Use With (Application) IgG Purification
Volume (Metric) 15 mL
Content And Storage Store in refrigerator (2–8°C).
Product Line POROS
Format Poros Bead
Stationary Phase Protein A
Purification Target IgG Antibodies, Ig Antibodies
Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.