Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ MabCapture™ G Select Resin

Catalog No. PI82083
Change view
Click to view available options
Volume (Metric):
5 mL
15 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PI82083 15 mL
PI82084 50 mL
PI82082 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PI82083 Supplier Thermo Scientific™ Supplier No. 82083
Add to Cart
Edge
Add to Cart
For Research Use Only. Not for use in diagnostic procedures.

MabCapture G Select antibody affinity purification resin provides high dynamic binding at low linear flow rates and maintains superior binding capacity at high linear flow rates

POROS™ MabCapture™ G Select Resin is a high-throughput, lower-priced Protein G chromatography resin manufactured using our own Protein G ligand to enable high-throughput purification performance. It is used in a variety of antibody affinity purification methods and is ideal for polyclonal IgG purification from mouse, human, cow, goat, and sheep serum, including human IgG3 and mouse IgG1 isotypes. POROS MabCapture G Select Resin demonstrates high dynamic binding performance at low linear flow rates and maintains superior binding capacity at high linear flow rates where other chromatography media products cannot operate.

  • Maintain high dynamic binding capacity at liner flow rates over 700 cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates, or both) due to the improvements made to the antibody binding efficiency
  • Obtain the highest dynamic binding capacity for antibodies vs. conventional agarose- or silica-based Protein G resins
  • Purify polyclonal IgG from mouse, human, cow, goat, and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Sanitize and depyrogenate with standard agents used for other chromatography products
  • Recognize major increases to process productivity and reduce the downstream bottleneck

Protein G is a bacterial cell wall protein isolated from group G Streptococci. Recombinant Protein G has been modified to eliminate its binding to albumin. The Protein G molecule contains two binding domains. Although the tertiary structures of Protein A and Protein G are very similar, their amino acid compositions differ significantly, resulting in different binding characteristics. Protein G has greater affinity than Protein A for most mammalian IgGs, especially for certain subclasses, including human IgG3, mouse IgG1, and rat IgG2a. Unlike Protein A, Protein G does not bind to human IgM, IgD, and IgA.

The development of humanized monoclonal antibodies has continued to fuel the growth of the biotherapeutic market. Addressing the therapeutic demand for monoclonal antibodies has led to major improvements to the cell-based expression systems that are used to manufacture these proteins. Consequently, these improvements have moved the processing bottleneck from upstream cell expression to downstream purification. The introduction of POROS MabCapture G Select chromatography resin was aimed at cracking open the downstream bottleneck and providing the capacity, efficiency, and flexibility needed to make improvements to the downstream purification platform, resulting in major increases to process productivity and antibody manufacturing.

TRUSTED_SUSTAINABILITY

Specifications

Type G Select Resin
Standard Components Protein G
For Use With (Application) IgG Purification
Quantity 15 ml
Volume (Metric) 15 mL
Content And Storage Store in refrigerator (2–8°C).
Product Line POROS
Format Poros Bead
Stationary Phase Protein G
Purification Target IgG Antibodies, Ig Antibodies
Shipping Condition Wet Ice
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.