Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ Oligo (dT)25 RoboColumn™ Pre-packed columns

Catalog No. A48350 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
A48350 600 μL
A48349 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A48350 Supplier Thermo Scientific™ Supplier No. A48350
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Thermo Scientific™ RoboColumn™ are pre-packed with POROS Oligo (dT)25 Affinity Resin, designed for the purification and isolation of mRNA from in vitro transcription (IVT) manufacturing processes.

These RoboColumn™ are pre-packed with POROS Oligo (dT)25 Affinity Resin, designed for the purification and isolation of mRNA from in vitro transcription (IVT) manufacturing processes. The resin effectively separates mRNA from components of the transcription reaction process, such as enzymes and plasmid DNA.

RoboColumn™ products are small chromatography columns that are provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.

POROS Oligo (dT)25 Affinity Resin (Cat. No. A47384) was developed to address the selectivity and capacity requirements for large-scale downstream purification of a range of mRNA constructs used in vaccine and gene therapy applications. Unlike alternate approaches, the resin selectively captures the mRNA via the polyadenylated (polyA) tail using simple salt and water purification steps.

Features of POROS Oligo (dT)25 Affinity Resin include:

  • Easy mRNA purification from crude transcription mix to effectively remove plasmid DNA and other IVT components
  • Simplified workflow helps to maximize efficiency, thereby reducing complexity of subsequent polish steps
  • Excellent scalability
  • Non-animal derived

POROS Oligo (dT)25 Affinity Resin allows for a robust platform and efficient purification process for a range of mRNA molecules and applications.

Oligo (dT) ligands are manufactured using a synthetic manufacturing process that are free of animal components.

Other POROS ion exchange resins, as well HIC and affinity resins, are also available in this RoboColumn format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Max. Pressure 8 bar
Pore Size Not Applicable
Stationary Phase Poly(dT) 25mer
Length (Metric) 3 cm
Quantity 1 Strip, 8 Columns
Particle Size 50 μm
For Use With (Equipment) Robotic Fluid Handlers
Type Affinity Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C.
Product Line POROS RoboColumn
Ligand Type Poly(dT) 25mer with Proprietary Linker
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Shipping Condition Wet Ice
Target mRNA
Column Format Pre-packed Robo Column
Bed Volume 600 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.