Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ XS Strong Cation Exchange Resin

Catalog No. pi82071
Change view
Click to view available options
Volume (Metric):
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PI82071 10 mL
PI82072 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PI82071 Supplier Thermo Scientific™ Supplier No. 82071
Only null left
Add to Cart
Edge
Add to Cart
For Research Use Only. Not for use in diagnostic procedures.

POROS XS is the first high-capacity, high-resolution resin that allows loading to >100 mg/mL in the presence of 15 mS/cm NaCl, while delivering unprecedented separation capability

POROS™ XS Strong Cation Exchange Resin is the first high-capacity, high-resolution resin that allows loading to more than 100 mg/mL capacity in the presence of up to 15 mS/cm NaCl, while delivering unprecedented separation capability. A major improvement on traditional cation exchangers, POROS XS resin sets the standard for cation exchange chromatography.

  • Dynamic binding capacity leads to reduced column size, smaller footprint, decreased buffer usage, and reduced cycling
  • High resolution supports better product yield and improved impurity clearance
  • Robust salt tolerance allows improved process flexibility, alignment, and process flow streamlining
  • Low back pressure and linear pressure responses help improve scalability
  • Rigid polymeric bead with covalent surface chemistry makes for easier handling and packing, and superior physical and chemical stability, resulting in a robust manufacturing process

Dynamic binding capacityPOROS XS resin has a high binding capacity of more than 100 mg/mL, at 5% breakthrough, over a broad range of process conditions.Higher dynamic binding capacity was achieved by optimizing the base bead pore structure and surface chemistry.

High resolution

Using an optimized 50 µm particle size, POROS XS resin maintains the superior resolving capacity of POROS™ HS resin while overcoming the challenging impurity levels associated with high titer processes. Resolution is consistent across a wide range of operating flow rates and is maintained independent of protein load.

Robust salt tolerance
POROS XS resin has high and consistent protein capacity across a broad range of salt concentrations up to 150 mM NaCl (15 mS/cm conductivity). The ability to load columns under higher conductivity reduces or helps eliminate the dilution of column feed streams or the removal of unit operations, such as tangential flow filtration. The optimized surface ionic capacity of POROS XS resin minimizes competitive binding between the protein of interest and the counterion, resulting in superior salt tolerance.

TRUSTED_SUSTAINABILITY

Specifications

Type XS Strong Cation Exchange Resin
Standard Components Crosslinked Poly(styrene-divinylbenzene)
For Use With (Application) Cation Exchange Chromatography
Quantity 10 mL
Volume (Metric) 10 mL
Content And Storage Store in refrigerator (2–8°C).
Product Line POROS
Format Poros Bead
Stationary Phase XS
Purification Target Protein
Shipping Condition Room Temperature
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.