Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ XQ RoboColumn™ Pre-packed columns

Catalog No. A30720 Shop All Thermo Scientific Products
Change view
Click to view available options
Column Capacity:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Column Capacity
A30720 600 μL
A30719 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30720 Supplier Thermo Scientific™ Supplier No. A30720
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XQ resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.

POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind and elute or flow-through modes of operation.

Features of the resin:

  • High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
  • High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
  • Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Flow Rate 16 to 1000 cm/hr.
Pore Size Not Applicable
Column Capacity 600 μL
Length (Metric) 3 cm
Quantity 1 Strip, 8 Columns
Particle Size 50 μm
For Use With (Equipment) Robotic Fluid Handlers
Type Anion Exchange Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 30°C. Do not freeze.
Product Line POROS
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Shipping Condition Ambient
Column Format Pre-packed Robo Column
Bed Volume 600 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.