Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ XQ Strong Anion Exchange Resin

Catalog No. p-6927056
Change view
Click to view available options
Volume (Metric):
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
PI82073 10 mL
PI82074 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. PI82073 Supplier Thermo Scientific™ Supplier No. 82073
Only null left
Add to Cart
Add to Cart
For Research Use Only. Not for use in diagnostic procedures.

POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that enables >140 mg/mL dynamic binding capacity while delivering exceptional separation performance

POROS™ XQ Strong Anion Exchange Resin is a next-generation, high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind & elute or flow-through modes of operation.

  • High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
  • High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
  • Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process

Dynamic binding capacity
POROS XQ resin has a high binding capacity of more than 140 mg/mL at 5% breakthrough, over a broad range of process conditions, with superior binding capacity across a broad range of pH and salt conditions. Higher dynamic binding capacity was achieved by optimizing the base bead pore structure and surface chemistry.

High resolution
Using an optimized 50 µm particle size, POROS XQ resin maintains the superior resolving capacity of the POROS resins, overcoming the challenging impurity levels associated with high titer processes. Resolution remains consistent across a wide range of operating linear flow rates.

Robust salt tolerance
POROS XQ resin has high and consistent protein capacity across a broad range of salt concentrations up to 140 mg/mL at 50 mM NaCl (6.6 mS/cm conductivity). The ability to load columns under higher conductivity reduces or helps eliminate the dilution of column feed streams or the removal of unit operations, such as tangential flow filtration.

TRUSTED_SUSTAINABILITY

Specifications

Type XQ Strong Anion Exchange Resin
Standard Components Quaternized Polyethyleneimine
For Use With (Application) Strong Anion Exchange Chromatography
Quantity 10 mL
Volume (Metric) 10 mL
Content And Storage Store in refrigerator (2–8°C).
Product Line POROS
Format Poros Bead
Stationary Phase Quaternized Polyethyleneimine
Purification Target Protein
Shipping Condition Room Temperature
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.